Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Carbonate, 99.99% Trace Metals Basis, Solstice

Catalog No. 60046676 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-676 50 g Poly Bottle
60-046-675 10 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-676 Supplier Solstice Supplier No. 37999950G

99.999% trace metals basis

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-87-6
Molecular Formula CH8N2O3
Molecular Weight (g/mol) 96.086
MDL Number MFCD00010890
InChI Key PRKQVKDSMLBJBJ-UHFFFAOYSA-N
Synonym ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate
PubChem CID 517111
IUPAC Name diazanium;carbonate
SMILES C(=O)([O-])[O-].[NH4+].[NH4+]

Specifications

Color White
pH 9 (at 100g/L 20°C)
Quantity 50 g
Linear Formula (NH4)2CO3
Beilstein 4253487
Solubility Information In water 320g/L at 20°C
Formula Weight 96.09g/mol
Percent Purity 99.999% (trace metals basis)
Odor Ammonia stinging
Packaging Poly Bottle
Appearance Solid
Vapor Pressure 69 hPa at 20°C
Decomposition Information Temperature : >160°C
Density 0.9g/cm3 at 20°C; Vapor Density: 2.7 (vs air)
Chemical Name or Material Ammonium carbonate
Show More Show Less
Hazard Statement P301 + P312 + P330
RTECSNumber 208-058-0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.