Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Carbonate Powder, Innovating Science™
SCI_ED

Catalog No. S000509 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000509 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning: Causes skin irritation, causes eye irritation, may cause respiratory irritation.
Catalog No. S000509 Supplier Aldon Corporation Supplier No. IS10077
Add to Cart
Add to Cart
Warning: Causes skin irritation, causes eye irritation, may cause respiratory irritation.

Chemical Identifiers

CAS 506-87-6
Molecular Weight (g/mol) 96.09 g/mol

Specifications

Physical Form Fine Crystals or Crystalline Powder
Quantity 500 g
Percent Purity 100%
Grade Laboratory
Packaging HDPE
Chemical Name or Material Ammonium Carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.