Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Cerium(Iv) Nitrate, ACS Reagent, ≥98.5%, Solstice

Catalog No. 60045239 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
50 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-239 50 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-045-239 Supplier Solstice Supplier No. 21547350G

missing translation for 'validMainframeMsg'

ACS Reagent,≥98.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16774-21-3
MDL Number MFCD00151121

Specifications

Melting Point 214°C
Color Orange
pH Acidic
Quantity 50 g
Linear Formula Ce(NH4)2(NO3)6
UN Number UN3085
Formula Weight 548.22g/mol
Percent Purity ≥98.5%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Powder/Crystals
Density 2.49g/cu.cm
Chemical Name or Material Ammonium Cerium(Iv) Nitrate
Show More Show Less
Hazard Statement P210-P220-P260-P280-P305 + P351 + P338-P370 + P378
Precautionary Statement H272-H290-H302-H314-H317-H410
DOTInformation Class 5.1 - PG 2 -EHS - acidic - Oxidizing solid, corrosive, n.o.s., HI: all
EINECSNumber 240-827-6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.