Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Cerium(Iv) Sulfate Dihydrate, ≥98.0% (RT), puriss. p.a. ACS Reagent, Solstice

Catalog No. 60045345 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-345 100 g Poly Bottle
60-045-346 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-045-345 Supplier Solstice Supplier No. 22269100G
Add to Cart
Add to Cart

Puriss. p.a., ACS Reagent,≥98.0% (RT)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10378-47-9
MDL Number MFCD00149417

Specifications

Quantity 100 g
Linear Formula Ce(NH4)4(SO4)4 · 2H2O
UN Number NONH for all modes of transport
Formula Weight 632.55g/mol
Percent Purity ≥98.0% (RT)
Grade ACS/puriss. p.a.
Packaging Poly Bottle
Chemical Name or Material Ammonium Cerium(Iv) Sulfate Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.