Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Chloride, 10% (w/v), Ricca Chemical
SDP

Catalog No. R06254004A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R06254004A 4 L Natural Poly Bottle
6254-16 500 mL Natural Poly Bottle
6254-32 1 L Natural Poly Bottle
R062540020F 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. R06254004A Supplier Ricca Chemical Company Supplier No. R06254004A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 12125-02-9 , 7732-18-5
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
PubChem CID 25517
ChEBI CHEBI:31206
IUPAC Name amine hydrochloride
SMILES N.Cl

Specifications

Color Colorless
CAS Min % 89.0, 9.0
CAS Max % 93.0, 11.0
Percent Purity 10%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Quantity 4 L
Physical Form Liquid
Concentration or Composition Notes 10% (w/v)
Chemical Name or Material Ammonium Chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.