Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Chloride-Ammonium Hydroxide, Buffer Solution, pH 10.0 ±0.1, For Water Hardness, APHA, Spectrum™ Chemical
SDP

Catalog No. 18600600 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-600-600 500 mL
18-600-602 1 L
18-600-601 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-600-600 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B205500MLGL
Add to Cart
Add to Cart

B-205, 12125-02-9

Spectrum™ Chemical Ammonium Chloride-Ammonium Hydroxide, Buffer Solution, pH 10.0 ± 0.1, For Water Hardness, APHA is an aqueous inorganic buffer solution concentrate that, when diluted, is used for the American Public Health Association's (APHA) method for determination of water hardness. Find the high quality buffer solution Ammonium Chloride-Ammonium Hydroxide at Spectrum. As an aqueous inorganic buffer solution concentrate, it is used for the American Public Health Association's (APHA) method for determination of water hardness after dilution.

Specifications

Chemical Name or Material Ammonium Chloride-Ammonium Hydroxide
CAS 1336-21-6, 12125-02-9, 7732-18-5
CAS Max % 1%, 98%
UN Number UN2672
Grade APHA/EPA
Packaging Amber Glass Bottle
pH 10 ±0.1
Quantity 500 mL
Physical Form Chemical Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.