Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Chloride, MP Biomedicals™

Catalog No. MP021914065 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021914065 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021914065 Supplier MP Biomedicals, Inc Supplier No. 02191406.5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used in research applications as an ACS reagent. Ammonium Chloride, MP Biomedicals is used in research to lyse human red blood cells, to study basic calcium phosphate crystals in fibroblasts, to isolate proteins from 50S ribosomal subunits of Bacillus ste

Industrial applications include as a flux for coating sheet iron with zinc; tinning; in dry and Leclanch batteries; dyeing, freezing mixtures, electroplating, to clean soldering irons, safety explosives, lustering cotton, tanning; in washing powders; manufacturing of dyes; in cement for iron pipes; for snow treatment (slows melting on ski slopes).

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
PubChem CID 25517
ChEBI CHEBI:31206
IUPAC Name amine hydrochloride
SMILES N.Cl

Specifications

Physical Form Powder
Quantity 500 g
Solubility Information Soluble in water (100mg/mL, clear, colorless solution). The presence of hydrochloric acid or sodium chloride decrease water solubility. Also soluble in methanol and ethanol.
Formula Weight 53.49
Percent Purity ≥99.5%
Grade ACS Reagent
Recommended Storage Room Temperature, (15-30°C).
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.