Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium chloride, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5%, Solstice

Catalog No. 60045903 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
6 x 500 g
6 x 1 kg
6 x 2.5 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-045-903 6 x 500 g
60-022-99 500 g
60-022-97 1 kg
60-022-98 2.5 kg
60-045-901 6 x 1 kg
60-045-902 6 x 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60-045-903 Supplier Solstice Supplier No. 311076X500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
PubChem CID 25517
ChEBI CHEBI:31206

Specifications

Quantity 6 x 500 g
Linear Formula NH4Cl
SMILES N.Cl
IUPAC Name azanium;chloride
Formula Weight 53.49
Percent Purity ≥99.5%
Grade ACS/puriss. p.a.
Vapor Pressure 1mmHg (160.4°C)
Density Vapor Density: 1.9 (vs air)
Chemical Name or Material Ammonium Chloride, Reag. Ph. Eur. reag. ISO
Hazard Statement P301 + P312 + P330-P305 + P351 + P338
Precautionary Statement H302-H319-H411
EINECSNumber 235-186-4
RTECSNumber BP4550000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.