Learn More
Specifications
Specifications
| Boiling Point | 520°C |
| Melting Point | 338°C |
| Color | White |
| pH | 4.0 to 5.5 (200g/L H2O 25°C) |
| Physical Form | Crystalline Powder |
| Quantity | 300 g |
| UN Number | 3077 |
| Formula Weight | 53.49 |
| Percent Purity | ≥99.5% (T) |
| Packaging | Plastic Bottle |
| Show More |
Safety and Handling
| EINECSNumber | (1)-0218 |
| RTECSNumber | BP4550000 |
| TSCA | Yes |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
