Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ammonium cobalt(II) phosphate, anhydrous, 98%

Click to view available options
Quantity:
50g
250g
14590-13-7
CoH4NO4P
171.942
MFCD02683100
FCXHHGTWMFGYPP-UHFFFAOYSA-L
ammonium cobaltous phosphate,ammonium cobalt ii phosphate,ammonium cobalt phosphate,kobaltammoniumphosphat,acmc-20alt0,ammonium cobalt orthophosphate,azanium cobalt 2+ phosphate,phosphoric acid, ammonium cobalt 2+ salt 1:1:1,ammonium cobalt ii phosphate, anhydrous,phosphoric acid,ammonium cobalt 2+ salt 1:1:1
167120
azanium;cobalt(2+);phosphate
[NH4+].[O-]P(=O)([O-])[O-].[Co+2]
14590-13-7
CoH4NO4P
171.942
MFCD02683100
FCXHHGTWMFGYPP-UHFFFAOYSA-L
ammonium cobaltous phosphate,ammonium cobalt ii phosphate,ammonium cobalt phosphate,kobaltammoniumphosphat,acmc-20alt0,ammonium cobalt orthophosphate,azanium cobalt 2+ phosphate,phosphoric acid, ammonium cobalt 2+ salt 1:1:1,ammonium cobalt ii phosphate, anhydrous,phosphoric acid,ammonium cobalt 2+ salt 1:1:1
167120
azanium;cobalt(2+);phosphate
[NH4+].[O-]P(=O)([O-])[O-].[Co+2]
Linear Formula NH4CoPO4
Merck Index 14,513
Formula Weight 171.94
Percent Purity 98%
Quantity 50g
Chemical Name or Material Ammonium cobalt(II) phosphate, anhydrous

GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P301+P310-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA