Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium copper(II) sulfate hexahydrate, 99%

Catalog No. AAA128340B
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA128340B 1000 g
AAA1283430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA128340B Supplier Thermo Scientific Chemicals Supplier No. A128340B
Add to Cart

CAS: 13587-26-3 | (NH4)2Cu(SO4)2·6H2O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13587-26-3
Molecular Formula (NH4)2Cu(SO4)2·6H2O
MDL Number MFCD00150440
Synonym Cupric ammonium sulfate

Specifications

Quantity 1000 g
Formula Weight 399.83 (291.75 Anhydrous)
Percent Purity 99%
Chemical Name or Material Ammonium copper(II) sulfate hexahydrate
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.