Ammonium dichromate, 99.5%, Alfa Aesar

Catalysts, source of pure nitrogen, in lithography

Manufacturer: Alfa Aesar 13444A3

Quantity 2kg

 View more versions of this product

Catalog No. AA13444A3

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7789-09-5
    Physical Form Crystalline
    Assay Percent Range 99.5%
    Molecular Formula (NH4)2Cr2O7
    Formula Weight 252.1
    Chemical Name or Material Ammonium dichromate

    Specifications

    Formula Weight 252.1
    Quantity 2kg
    Physical Form Crystalline
    Assay Percent Range 99.5%
    Molecular Formula (NH4)2Cr2O7
    CAS 7789-09-5
    Chemical Name or Material Ammonium dichromate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit