Learn More
Ammonium fluoride, 96%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Ammonium fluoride is used for etching of glass, preservation of food, in printing textile, and as mothproofing agent. It is used in chemical analysis and in brewing. It is used in the synthesis of NaHoF4 and NaEuF4, which are important in solid state lasers as well as photoluminescence. The fluoride ion is used as a base and nucleophile in organic synthesis. Ammonium fluoride providee safe deprotection strategy for the tert-butyldimethylsilyl (TBS) group during RNA synthesis.
Solubility
Freely soluble in water. Slightly soluble in alcohol.
Notes
Hygroscopic and non-cumbustible. On heating, it sublimes and decomposes to hydrogen fluoride and ammonia gas, which can recombine to form ammonium fluoride.
Specifications
Specifications
| Quantity | 2500 g |
| Linear Formula | NH4F |
| UN Number | UN2505 |
| Merck Index | 14,522 |
| Solubility Information | Freely soluble in water. Slightly soluble in alcohol. |
| Formula Weight | 37.04 |
| Percent Purity | 96% |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Density | 1.01 |
| Show More |
Safety and Handling
| Hazard Category | H301+H311+H331 |
| Hazard Statement | GHS H Statement H301-H311-H331 Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: AMMONIUM FLUORIDE |
| EINECSNumber | 235-185-9 |
| RTECSNumber | BQ6300000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.