Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Formate, for HPLC, Solstice

Catalog No. 6002035 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-020-35 250 g
60-020-36 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-020-35 Supplier Solstice Supplier No. 17843250G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

for HPLC,≥99.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-69-2
Molecular Formula CH5NO2
Molecular Weight (g/mol) 63.056
MDL Number MFCD00013103
InChI Key VZTDIZULWFCMLS-UHFFFAOYSA-N
Synonym ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny
PubChem CID 2723923
ChEBI CHEBI:63050
IUPAC Name azanium;formate
SMILES C(=O)[O-].[NH4+]

Specifications

Quantity 250 g
Linear Formula HCO2NH4
UN Number NONH for all modes of transport
Beilstein 3625095
Solubility Information Soluble in water 0.1g/mL at 25°C
Formula Weight 63.06g/mol
Percent Purity ≥99%
Density 1.26g/mL at 25°C
Chemical Name or Material Ammonium Formate
EINECSNumber 208-753-9
RTECSNumber BQ6650000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.