Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium hexabromoosmate(IV), 99.9% (metals basis), Os 26.5 % min

Catalog No. p-7049069
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1264803 1 g
AA1264806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1264803 Supplier Thermo Scientific Chemicals Supplier No. 01264803
Only null left
Add to Cart

CAS: 24598-62-7 | Br6H8N2Os | 705.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24598-62-7
Molecular Formula Br6H8N2Os
Molecular Weight (g/mol) 705.73
MDL Number MFCD00049389
InChI Key ZPHFWWAVHILXPL-UHFFFAOYSA-J
Synonym diammonium hexabromoosmiumdiuide, osmate 2-,hexabromo-,ammonium 1:2 , oc-6-11
PubChem CID 11136267
IUPAC Name diazanium;hexabromoosmium(2-)
SMILES [NH4+].[NH4+].[Br-].[Br-].[Br-].[Br-].[Br-].[Br-].[Os+4]

Specifications

Color Black
Physical Form Powder
Quantity 1 g
Formula Weight 705.73
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Os 26.5 % min.
Odor Odorless
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Ammonium hexabromoosmate(IV)
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 246-340-5
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.