Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium hexafluorogermanate, 99.99% min (metals basis)

Catalog No. p-7058752
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4317906 5 g
AA4317914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4317906 Supplier Thermo Scientific Chemicals Supplier No. 04317906
Only null left
Add to Cart

CAS: 16962-47-3 | H8F6GeN2

Ammonium hexafluorogermanate is used to substitutes Al with Ge in Y type zeolites.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16962-47-3
Molecular Formula H8F6GeN2
MDL Number MFCD00010880

Specifications

Color White
Physical Form Crystalline
Quantity 5 g
Linear Formula (NH4)2GeF6
UN Number UN3288
Solubility Information Soluble in water.
Formula Weight 222.71
Percent Purity ≥99.99%
Density 2.564 g/mL
Assay (metals basis)
Chemical Name or Material Ammonium hexafluorogermanate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 241-037-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.