Alfa Aesar™ Ammonium hexafluorosilicate, 98%

Etching glass, in soldering flux

Manufacturer: Alfa Aesar™ 6910030

Quantity 250g

 View more versions of this product

Catalog No. AA6910030

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16919-19-0
    Molecular Formula (NH4)2SiF6
    Formula Weight 178.14
    MDL Number MFCD00010887
    UN Number UN2854
    Merck Index 14,527
    Chemical Name or Material Ammonium hexafluorosilicate

    Specifications

    Formula Weight 178.14
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula (NH4)2SiF6
    CAS 16919-19-0
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H331
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    MDL Number MFCD00010887
    UN Number UN2854
    TSCA TSCA
    Physical Form Powder
    Merck Index 14,527
    Quantity 250g
    Solubility Information Slightly soluble in water; Insoluble in alcohol, acetone
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: AMMONIUM FLUOROSILICATE
    Chemical Name or Material Ammonium hexafluorosilicate
    Assay 98%

    GHS H Statement
    H301-H311-H331
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.

    GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit