Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide, 10% (w/v) NH3 Solution, (U.S.P. Test Solution), Spectrum™ Chemical
SDP

Catalog No. 18600603 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-600-603 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-600-603 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A158500MLPL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

A-158, 1336-21-6

Spectrum™ Chemical Ammonium Hydroxide, 10 Percent (w/v) NH3 Solution, (U.S.P. Test Solution) is a solution of ammonia in water. Aqueous ammonia, as its otherwise known is used as a complexant and base in qualitative inorganic analysis. It becomes a deep blue color with copper solutions. Used to dissolve silver oxide residues, such as the ones formed from Tollens reagent, it is also commonly used in numerous cleaning agents, such as window cleaning formulas as well as a complexant that is manufactured to parts-per-billion trace impurities and has been used in the semiconductor industry to dissolve heavy or precious metal components.

Chemical Identifiers

CAS 1336-21-6 , 7732-18-5
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
IUPAC Name amine hydrate
SMILES N.O

Specifications

CAS Max % 0.1%, 99.9%
UN Number UN2672
Percent Purity 10% ±0.3
Grade USP
Packaging Poly Bottle
pH 10.6 to 11.6
Quantity 500 mL
Physical Form Chemical Solution
Chemical Name or Material Ammonium Hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.