Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide, 25% (v/v), Ricca Chemical
SDP

Catalog No. 63732 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
63732 1 L Natural Poly Bottle
50126832 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 63732 Supplier Ricca Chemical Company Supplier No. 63732
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1336-21-6
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

pH Alkaline
Color Colorless
Assay Percent Range 25% (v/v) Aqueous Solution (1 + 3)
Solubility Information Solubility in Water: Infinite
Quantity 1 L
Percent Purity 24.3 to 25.7% v/v
Grade Laboratory
Odor Ammonia
Packaging Natural Poly Bottle
Physical Form Liquid
Chemical Name or Material Ammonium Hydroxide Solution
Show More Show Less
Recommended Storage Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.