Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide, 30%, BAKER INSTRA-ANALYZED Reagent, J.T. Baker™

Catalog No. 14650244 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Poly container
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
14-650-244 500 mL Poly container
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-650-244 Supplier Avantor Supplier No. 973301

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name azanium;hydroxide
SMILES N.O

Specifications

Assay Percent Range 28.0 to 30.0 %
Quantity 500 mL
Formula Weight 35.05
Percent Purity 28.0 to 30.0%
Packaging Poly container
Chemical Name or Material Ammonium Hydroxide, 30%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.