Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide, 50% (v/v), Ricca Chemical
SDP

Catalog No. R064000055E Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
16 gal.
200 L
55 gal.
Packaging:
Cubitainer
Natural Poly Bottle
Poly Drum
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R064000055E 55 gal. Poly Drum
640-16 500 mL Natural Poly Bottle
640-32 1 L Natural Poly Bottle
R06400004F 4 L Cubitainer
640-1 4 L Natural Poly Bottle
R064000020F 20 L Cubitainer
R0640000200 200 L Poly Drum
64016G 16 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. R064000055E Supplier Ricca Chemical Company Supplier No. R064000055E
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1336-21-6 , 7732-18-5
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name amine hydrate
SMILES N.O

Specifications

Color Colorless
CAS Min % 49.0
CAS Max % 51.0
Concentration or Composition Notes 50% (v/v)
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
Quantity 55 gal.
Percent Purity 48.5 to 51.5% v/v
Grade Laboratory
Packaging Poly Drum
Solvent or Matrix Aqueous Solution
Physical Form Liquid
Chemical Name or Material Ammonium Hydroxide
Show More Show Less
ShelfLife 24 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.