Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide, 50% (v/v) Solution, Spectrum™ Chemical
SDP

Catalog No. 18608683 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-683 1 L Poly Bottle
18-608-691 500 mL Poly Bottle
18-608-682 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-608-683 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A1601LT
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1336-21-6 , 7732-18-5

Specifications

CAS Max % 1
Concentration 50%
Quantity 1 L
Grade Chemical Solution
Packaging Poly Bottle
Chemical Name or Material Ammonium Hydroxide Solution

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.