Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide ACS Grade, Innovating Science™
SCI_ED

Catalog No. S000513 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000513 2.5 L
S000512 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger: Causes severe skin burns and eye damage, may cause respiratory irritation, very toxic to aquatic life.
Catalog No. S000513 Supplier Aldon Corporation Supplier No. IS10088
Add to Cart
Add to Cart
Danger: Causes severe skin burns and eye damage, may cause respiratory irritation, very toxic to aquatic life.

Chemical Identifiers

CAS 7732-18-5 , 1336-21-6

Specifications

Concentration 14.8 M
Quantity 2.5 L
Grade ACS
Packaging Amber Glass
Chemical Name or Material Ammonium Hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.