Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ammonium Hydroxide Solution, 14.8M
SCI_ED

Catalog No. s25159 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Reagent Grade
Reagent Grade - Safe-Cote
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25159 500 mL Reagent Grade - Safe-Cote
S25159A 2.5 L Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25159
Only null left
Add to Cart
Edge
Add to Cart

Ammonium Hydroxide Solution for use in science classrooms and labs.

  • Reagent grade solution suitable for lab use
  • Comes in 500 mL and 2.5 L volumes

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name amine hydrate
SMILES N.O

Specifications

Color Colorless
Assay Percent Range < 30% w/v
Linear Formula NH4OH
Density ∽0.9 to 1.0g/mL
Quantity 500 mL
Formula Weight 35.04
Grade Reagent
Packaging Reagent Grade - Safe-Cote
Chemical Name or Material Ammonium Hydroxide Solution
Safety handling Check Alert storage code Red
ShelfLife 5 years
DOTInformation DOT Class 8, III, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.