Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide Solution, ACS Reagent, 28.0 to 30.0% NH3 basis, Solstice

Catalog No. 6000310 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
6 x 500 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-003-10 6 x 500 mL
60-003-04 100 mL
60-003-09 500 mL
60-003-06 1 L
60-003-07 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-003-10 Supplier Solstice Supplier No. 2212286X500ML/DEL

ACS Reagent, 28.0-30.0% NH3 basis

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
SMILES N.O

Specifications

Linear Formula NH4OH
Concentration 28 to 30% NH3 basis
Beilstein 3587154
Density 0.9g/mL at 25°C
Quantity 6 x 500 mL
IUPAC Name azanium;hydroxide
Explosion Limit 0.27
Formula Weight 35.05g/mol
Grade ACS Reagent
Packaging Glass Bottle
Residue after Ignition ≤0.002%
Chemical Name or Material Ammonium Hydroxide Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.