Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide Solution, puriss. p.a., 0%, Solstice

Catalog No. 60044624 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-624 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-044-624 Supplier Solstice Supplier No. 098611L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss. p.a., packed in coated, shock- and leak-protected glass bottle, 25% NH3 in H2O (T)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name azanium;hydroxide
SMILES N.O

Specifications

pH Alkaline
Color Colorless
Boiling Point 24°C (1013hPa)
Melting Point -92°C
Linear Formula NH4OH
UN Number UN2672
Beilstein 3587154
Density 0.91g/mL at 20°C, 0.9g/mL at 25°C
Solubility Information Completely miscible
Quantity 1 L
Explosion Limit 0.27
Formula Weight 35.05g/mol
Percent Purity 0%
Grade puriss. p.a.
Odor Stinging ammoniacal
Packaging Glass bottle
Physical Form Liquid
Chemical Name or Material Ammonium Hydroxide Solution
Show More Show Less
Hazard Statement P273-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P391
Precautionary Statement H302-H314-H335-H400
DOTInformation 8/PGIII
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.