Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide Solution, TraceSELECT™, Solstice

Catalog No. 60044623 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-623 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-044-623 Supplier Solstice Supplier No. 09857500ML
Add to Cart
Add to Cart

TraceSELECT™, for trace analysis,≥25% in H2O

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
SMILES N.O

Specifications

Color Colorless
Boiling Point 24°C
Melting Point -92°C
Linear Formula NH4OH
Concentration ≥25% in H2O
UN Number UN2672
Beilstein 3587154
Density 0.9g/mL at 25°C, 0.91g/mL at 20°C
Quantity 500 mL
IUPAC Name amine hydrate
Explosion Limit 0.27
Formula Weight 35.05g/mol
Grade TraceSELECT
Packaging Poly Bottle
Vapor Pressure 1900 hPa at 50°C, 837 hPa at 20°C
Physical Form Liquid
Chemical Name or Material Ammonium Hydroxide Solution
Show More Show Less
Hazard Statement P273-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P391
Precautionary Statement H302-H314-H335-H400
DOTInformation 8/PGIII
Recommended Storage 2° to 8°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.