Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium hydroxide solution, TraceSELECT Ultra, for ultratrace analysis,≥20% in H2O, Solstice

Catalog No. 60044947 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-044-947 1 L
60-044-948 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-044-947 Supplier Solstice Supplier No. 167481L
Add to Cart
Add to Cart

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5 , 1336-21-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
MDL Number MFCD00066650
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Ammonia aqueous, Ammonia water
IUPAC Name water
SMILES O

Specifications

Boiling Point 24°C
CAS Min % ≥70.00, ≥20.00
CAS Max % <90.00, <30.00
Beilstein 3587154
Density 0.90 g/cm3
Quantity 1 L
Grade Trace Analysis
Packaging Poly Bottle
Physical Form Liquid
Hazard Category Danger
Hazard Statement H314+H335
Precautionary Statement P261+P264+P271+P280
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.