Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium iron(III) citrate, 16.5 to 18.5% Fe

Catalog No. p-4509959
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC211842500 250 g Plastic bottle
AC211840010 1 kg Plastic bottle
AC211840025 2.5 kg Plastic bottle
AC211840100 10 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC211842500 Supplier Thermo Scientific Chemicals Supplier No. 211842500
Only null left
Add to Cart

CAS: 1185-57-5 | C6H11FeNO7 | 265 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185-57-5
Molecular Formula C6H11FeNO7
Molecular Weight (g/mol) 265
MDL Number MFCD00013099
InChI Key FRHBOQMZUOWXQL-UHFFFAOYSA-N
Synonym ammonium ferric citrate, ferri seltz, ammonii ferri citras, ferric ammonium citrate, iron ammonium citrate, ammonium iron 3+ citrate, ammonium iron iii citrate, iron ammonium citrate, green, ferri-ammoniumcitrat, braunes, ferriseltz
PubChem CID 118984355
IUPAC Name azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+)
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3]

Specifications

Color Brown to Brown-Yellow
Physical Form Powder
Quantity 250 g
Merck Index 15, 4044
Solubility Information Solubility in water: 1200g/L (20 c). Other solubilities: practically insoluble in alcohol
Formula Weight 265
Percent Purity 16.5 to 18.5% (Fe)
Infrared Spectrum Authentic
Packaging Plastic bottle
Chemical Name or Material Ammonium iron(III) citrate, 16.5 to 18.5% Fe
Hazard Category HYGROSCOPIC
Hazard Statement LIGHT SENSITIVE
EINECSNumber 214-686-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.