Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium iron(III) sulfate dodecahydrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., 99.0-102.0%, crystalline, slightly violet, Solstice

Catalog No. 6002300 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-023-00 2.5 kg
60-023-01 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-023-00 Supplier Solstice Supplier No. 311112.5KG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., 99.0-102.0%, crystalline, slightly violet

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-83-7
Molecular Formula FeH28NO20S2
Molecular Weight (g/mol) 482.176
InChI Key LCPUDZUWZDSKMX-UHFFFAOYSA-K
Synonym ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
PubChem CID 197097
IUPAC Name azanium;iron(3+);disulfate;dodecahydrate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]

Specifications

Color Violet
Physical Form Crystalline
Quantity 2.5 kg
Linear Formula NH4Fe(SO4)2 · 12H2O
MDL Number MFCD00150004
UN Number NONH for all modes of transport
Solubility Information In accordance with water, 10g/dL
Formula Weight 482.19g/mol
Percent Purity 99 to 102%
Grade ACS/puriss. p.a.
Chemical Name or Material Ammonium Iron(Iii) Sulfate Dodecahydrate, Reag. Ph. Eur. reag. ISO
Show More Show Less
EINECSNumber 233-382-4
RTECSNumber WS5900000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.