Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Metavanadate, ACS Reagent, ≥99%, Solstice

Catalog No. 60046765 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-765 50 g Poly Bottle
60-046-764 250 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-765 Supplier Solstice Supplier No. 39812850G

missing translation for 'validMainframeMsg'

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7803-55-6
Molecular Formula H4NO3V
Molecular Weight (g/mol) 116.98
MDL Number MFCD00011430
InChI Key YBVKNHXQSRDWAA-UHFFFAOYSA-M
Synonym ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate
PubChem CID 516859
IUPAC Name azanium;oxido(dioxo)vanadium
SMILES N.O[V](=O)=O

Specifications

Melting Point 200°C
Color Yellow
pH 7
Quantity 50 g
Linear Formula NH4VO3
UN Number UN2859
Solubility Information In water 5.1g/L at 20°C
Formula Weight 116.98g/mol
Percent Purity ≥99%
Concentration or Composition (by Analyte or Components) 5.1g/L
Odor Odorless
Packaging Poly Bottle
Appearance Powder
Density 2.32g/cu.cm at 25°C
Chemical Name or Material Ammonium metavanadate
Show More Show Less
Hazard Statement P260-P284-P301 + P310-P305 + P351 + P338-P310
DOTInformation 6.1/PGII
RTECSNumber 232-261-3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.