Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Metavanadate, ACS Reagent, Reagents

Catalog No. 01338075
Change view
Click to view available options
:
Quantity: 100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-338-075 Quantity: 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-338-075 Supplier Reagents Holdings Llc Supplier No. C1139600100C2
Only null left
Add to Cart
Add to Cart

Ammonium Metavanadate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7803-55-6
Molecular Formula H4NO3V
Molecular Weight (g/mol) 116.98
MDL Number MFCD00011430
InChI Key YBVKNHXQSRDWAA-UHFFFAOYSA-M
Synonym Ammonium Vanadate
IUPAC Name vanadiumoylol amine
SMILES N.O[V](=O)=O

Specifications

Melting Point 200°C
Color White to Pale Yellow
Physical Form Solid
Quantity 100 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 2.326
Chemical Name or Material Ammonium Metavanadate, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.