Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Metavanadate, Solstice

Catalog No. 6002049 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-020-49 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-020-49 Supplier Solstice Supplier No. 2055591KG

missing translation for 'validMainframeMsg'

0.99

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7803-55-6
Molecular Formula H4NO3V
Molecular Weight (g/mol) 116.98
MDL Number MFCD00011430
InChI Key YBVKNHXQSRDWAA-UHFFFAOYSA-M
Synonym ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate
PubChem CID 516859
SMILES N.O[V](=O)=O

Specifications

Quantity 1 kg
Linear Formula NH4VO3
UN Number 2859
IUPAC Name azanium;oxido(dioxo)vanadium
Formula Weight 116.98g/mol
Percent Purity 99%
Density 2.32g/cm3at 25°C
Chemical Name or Material Ammonium Metavanadate
Hazard Statement P260-P284-P301 + P310-P305 + P351 + P338-P310
Precautionary Statement H301-H315-H319-H330-H335
EINECSNumber 232-261-3
RTECSNumber YW0875000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.