Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Molybdate Tetrahydrate, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RDCA0450500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCA0450500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCA0450500 Supplier Ricca Chemical Company Supplier No. RDCA0450500B1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 12054-85-2
Molecular Weight (g/mol) 1235.86 g/mol

Specifications

Color Green-Yellow to White
CAS Min % 98.0
CAS Max % 100.0
Physical Form Crystals
Quantity 500 g
Percent Purity 81.0 to 83.0%
Grade ACS
Concentration or Composition (by Analyte or Components) 4-Hydrate
Packaging White Poly Wide Mouth Bottle
Chemical Name or Material Ammonium Molybdate
ShelfLife 60 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.