Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Molybdate, For Phosphorus, Certified, 4% (w/v), LabChem™

Catalog No. LC112101 Shop All LabChem Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC112101 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC112101 Supplier LabChem, Inc. Supplier No. LC112101
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 12054-85-2 , 7732-18-5

Specifications

Color Colorless
CAS Max % 96, 4
Linear Formula (NH4)6Mo7O24·4H2O
Density 1g/mL
Solubility Information Soluble in water
Formula Weight 1235.86
Percent Purity 4% (w/v)
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Suitability Passes Test
Decomposition Information Gaseous Ammonia
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Ammonium Molybdate, For Phosphorus
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.