Learn More
Ammonium Molybdate Tetrahydrate, ACS Reagent, Reagents
Ammonium Molybdate Tetrahydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
$150.50
Chemical Identifiers
| CAS | 12054-85-2 |
|---|---|
| Molecular Formula | H6MoNO2 |
| Molecular Weight (g/mol) | 148.00 |
| InChI Key | XCOMAVCHADVANQ-UHFFFAOYSA-O |
| Synonym | Ammonium molybdenum oxide |
| IUPAC Name | ammonium hydrate molybdenum oxidandiide |
| SMILES | [NH4+].O.[O--].[Mo] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
01-338-100
|
Reagents Holdings Llc
C1135000100C2 |
100 g |
Each of 1 for $150.50
|
|
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Chemical Identifiers
| 12054-85-2 | |
| 148.00 | |
| Ammonium molybdenum oxide | |
| [NH4+].O.[O--].[Mo] |
| H6MoNO2 | |
| XCOMAVCHADVANQ-UHFFFAOYSA-O | |
| ammonium hydrate molybdenum oxidandiide |
Specifications
| White to Slightly Yellow-Green | |
| 190°C | |
| H6MoNO2 | |
| XCOMAVCHADVANQ-UHFFFAOYSA-O | |
| ammonium hydrate molybdenum oxidandiide | |
| ACS | |
| 2.498 | |
| Solid |
| 12054-85-2 | |
| 190°C | |
| Ammonium molybdenum oxide | |
| [NH4+].O.[O--].[Mo] | |
| 148.00 | |
| Glass Bottle | |
| Non-sterile | |
| Ammonium Molybdate Tetrahydrate, ACS Reagent |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.