Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Molybdate Tetrahydrate (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. A674500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
3 kg
10 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A674-500 500 g Poly Bottle
A674-3 3 kg Poly Bottle
A674-10 10 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A674-500
Only null left
Add to Cart

Chemical Identifiers

CAS 12054-85-2
Molecular Formula H24MO7N6O24 · 4 H2O
MDL Number MFCD00167059
Synonym Ammonium Heptamolybdate Tetrahydrate

Specifications

Melting Point 190°C
Color White
pH 5.0 to 5.5
Physical Form Solid
Quantity 500 g
Assay Percent Range 81.0 to 83.0%
Linear Formula (NH4)6Mo7O24·4H2O
Formula Weight 1235.86
Percent Purity 81.0 to 83.0%
Grade Certified ACS
Identification Pass Test
Packaging Poly Bottle
Decomposition Information >150°C
Density 2.490g/cm³
Chemical Name or Material Ammonium Molybdate Tetrahydrate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Harmful if swallowed. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Obtain medical attention. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

NFPA

Health:2
Flammability:1
Instability:0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.