Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Molybdate, Tetrahydrate, MP Biomedicals™

Catalog No. MP215249791 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215249791 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215249791 Supplier MP Biomedicals, Inc Supplier No. 0215249791
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ammonium Molybdate, Tetrahydrate, MP Biomedicals is an analgesic.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12054-85-2
Molecular Formula (NH4)6Mo7O24.4H2O
Synonym Ammonium heptamolybdate tetrahydrate, Molybdic acid ammonium salt tetrahydrate

Specifications

Physical Form Crystalline Powder
Quantity 1 kg
Solubility Information Soluble in water (1g/mL).
Formula Weight 1235.86
Grade ACS
Chemical Name or Material Ammonium Molybdate Tetrahydrate
Recommended Storage Room Temperature (15–30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.