Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Molybdate Tetrahydrate, puriss. p.a. ACS Reagent, Solstice

Catalog No. 6001883 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
25 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-018-83 500 g
60-018-80 100 g
60-018-81 1 kg
60-018-82 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-018-83 Supplier Solstice Supplier No. 09880500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss. p.a., ACS reagent,≥99.0% (T)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12054-85-2
MDL Number MFCD00167059

Specifications

Physical Form Solid
Quantity 500 g
Linear Formula (NH4)6Mo7O24 · 4H2O
UN Number NONH for all modes of transport
Formula Weight 1235.86g/mol
Percent Purity ≥99% (T)
Grade ACS/puriss. p.a.
Packaging Plastic Bottle
Density 2.498g/mL at 25°C
Chemical Name or Material Ammonium Molybdate Tetrahydrate
EINECSNumber 234-320-9
RTECSNumber QA5076000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.