Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Molybdate TS for Mannitol, Reagents

Ammonium Molybdate TS for Mannitol, Meets USP specifications, for general laboratory use. Manufactured in ISO 9001 facility.

Supplier:  Reagents Holdings Llc CA0060001A

Quantity 1 L

Catalog No. 01-336-123


Only null left
Add to Cart

Description

Description

Specifications

Chemical Identifiers

7732-18-5, 12054-85-2
18.02
water
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
O

Specifications

Colorless to Light Yellow
100°C
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
USP
1.1
1 L
Liquid
7732-18-5, 12054-85-2
H2O
O
18.02
Poly Bottle
5 to 6
Non-sterile
Ammonium Molybdate TS for Mannitol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.