Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Nitrate, Solstice

Catalog No. 6000352 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-003-52 500 g
60-003-50 1 kg
60-003-51 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-003-52 Supplier Solstice Supplier No. 31114500G

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥98%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6484-52-2
Molecular Formula H4N2O3
Molecular Weight (g/mol) 80.04
MDL Number MFCD00011425
InChI Key DVARTQFDIMZBAA-UHFFFAOYSA-O
Synonym ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills
PubChem CID 22985
ChEBI CHEBI:63038
IUPAC Name azanium;nitrate
SMILES [NH4+].[O-][N+]([O-])=O

Specifications

Quantity 500 g
Linear Formula NH4NO3
UN Number 1942
Formula Weight 80.04g/mol
Percent Purity ≥98%
Grade ACS/puriss. p.a.
Packaging Plastic Bottle
Residue after Ignition ≤0.01% (as SO4)
Chemical Name or Material Ammonium Nitrate, Reag. Ph. Eur. reag. ISO
Hazard Statement P220-P261-P305 + P351 + P338
Precautionary Statement H272-H315-H319-H335
EINECSNumber 229-347-8
RTECSNumber BR9050000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.