Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ammonium Oxalate, Monohydrate
SCI_ED

Catalog No. S25174 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Lab Grade - Crystal
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25174 100 g Lab Grade - Crystal
S25174A 500 g Lab Grade - Crystal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S25174
Add to Cart
Add to Cart
This item is not returnable. View return policy

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 6009-70-7
Molecular Formula C2H10N2O5
Molecular Weight (g/mol) 142.111
InChI Key MSMNVXKYCPHLLN-UHFFFAOYSA-N
Synonym ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate
PubChem CID 516808
IUPAC Name diazanium;oxalate;hydrate
SMILES C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O

Specifications

Color White
Quantity 100 g
Assay Percent Range >99% w/v
Linear Formula (NH4)2C2O4·H2O
Formula Weight 142.11
Grade Laboratory
Packaging Lab Grade - Crystal
Density 1.5g/mL
Chemical Name or Material Ammonium Oxalate, Monohydrate
Safety handling Check Alert storage code Red
ShelfLife 5 years
DOTInformation DOT Class 6.1, III, : Poison
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.