Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ammonium Phosphate, Dibasic
SCI_ED

Catalog No. S25175A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Reagent Grade
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25175A 100 g Reagent Grade
S25175 500 g Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25175A

missing translation for 'validMainframeMsg'

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7783-28-0
Molecular Formula H9N2O4P
Molecular Weight (g/mol) 132.06
MDL Number MFCD00010891
InChI Key MNNHAPBLZZVQHP-UHFFFAOYSA-N
Synonym diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
PubChem CID 24540
ChEBI CHEBI:63051
IUPAC Name phosphoric acid diamine
SMILES N.N.OP(O)(O)=O

Specifications

Color White
Quantity 100 g
Linear Formula (NH4)2HPO4
Formula Weight 132.06
Grade Reagent
Packaging Reagent Grade
Density 1.619g/mL
Chemical Name or Material Ammonium Phosphate, Dibasic
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.