Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Phosphate Dibasic (Colorless Crystals), Fisher BioReagents

Catalog No. BP361500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP361-500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP361-500

CAS: 7783-28-0 | H9N2O4P | 132.06 g/mol

Poly Bottle; 500g

Ammonium Phosphate Dibasic is suitable for use in many biological applications.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ammonium Phosphate Dibasic
Color White
CAS 7783-28-0
pH 7.9 to 8.3
Physical Form Crystalline Solid
Quantity 500 g
Assay Percent Range 98.0 to 101.0 %
Molecular Formula H9N2O4P
Linear Formula (NH4)2HPO4
MDL Number MFCD00010891
Merck Index 15, 540
Synonym diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
InChI Key MNNHAPBLZZVQHP-UHFFFAOYSA-N
SMILES N.N.OP(O)(O)=O
IUPAC Name phosphoric acid diamine
Molecular Weight (g/mol) 132.06
PubChem CID 24540
ChEBI CHEBI:63051
Formula Weight 132.06
Percent Purity 98.0 to 101.0%
Identification Pass Test
Packaging Poly Bottle
Density 1.619g/cm³
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.