Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Phosphate Dibasic, FCC, 96-102%, Spectrum™ Chemical
SDP

Catalog No. 18600640 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-640 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-600-640 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. AM184500GM

missing translation for 'validMainframeMsg'

AM184, 7783-28-0, (NH4)2HPO4

Spectrum™ Chemical Ammonium Phosphate Dibasic, FCC is a food ingredient used as an acidity regulator and raising agent. It is listed on the FDA's Generally Recognized As Safe (GRAS) list of substances. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7783-28-0
Molecular Formula H9N2O4P
Molecular Weight (g/mol) 132.06
MDL Number MFCD00010891
InChI Key MNNHAPBLZZVQHP-UHFFFAOYSA-N
IUPAC Name phosphoric acid diamine
SMILES N.N.OP(O)(O)=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 96 to 102%
Grade FCC
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.