Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Phosphate Solution, 400 g (NH4)2HPO4 + 800 mL Water, Ricca Chemical
SDP

Catalog No. 69016 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
690-16 500 mL Natural Poly Bottle
6904 120 mL Natural Poly Bottle
R06900001A 1 L Natural Poly Bottle
R06900004A 4 L Natural Poly Bottle
R069000020F 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 690-16 Supplier Ricca Chemical Company Supplier No. 69016
Add to Cart
Add to Cart

APHA, EPA for Sulfide Testing

Chemical Identifiers

Molecular Formula H9N2O4P
Molecular Weight (g/mol) 132.06
MDL Number MFCD00010891
InChI Key MNNHAPBLZZVQHP-UHFFFAOYSA-N
IUPAC Name phosphoric acid diamine
SMILES N.N.OP(O)(O)=O

Specifications

CAS 7732-18-5
Grade Laboratory
Packaging Natural Poly Bottle
pH ∼8
Quantity 500 mL
Chemical Name or Material AMMONIUM PHOSPHATE, 400 + 800
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.