Learn More
Ammonium Standard, Inorganic, Quantitative, 125 mL
Specifications
Specifications
| CAS Max % | 99.9, 0.1 |
| Analyte or Component Names | Water,Ammonium |
| Element | Ammonium |
| Solvent or Matrix | Water |
| Quantity | 125 mL |
| Concentration or Composition (by Analyte or Components) | 1000μg/mL |
| Chemical Name or Material | Ammonium Standard |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.