Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Standard Stock Solution (250 ppm NH4), Ricca Chemical
SDP

Catalog No. 6914 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
691-4 120 mL Natural Poly Bottle
691-16 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 691-4 Supplier Ricca Chemical Company Supplier No. 6914
Add to Cart
Add to Cart

EP Standard Solution

Chemical Identifiers

CAS 12125-02-9 , 7732-18-5
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
PubChem CID 25517
ChEBI CHEBI:31206
IUPAC Name amine hydrochloride
SMILES N.Cl

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 250ppm NH4
CAS Min % 98.0, 0.07
CAS Max % 100.0, 0.08
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Quantity 120 mL
Chemical Name or Material Ammonium Standard Stock Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.