Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Sulfate, 3.5 Molar, Reagents

Catalog No. 01335956
Change view
Click to view available options
:
Quantity: 20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-335-956 Quantity: 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-956 Supplier Reagents Holdings Llc Supplier No. CA10120020F
Only null left
Add to Cart
Add to Cart

Ammonium Sulfate, 3.5 Molar, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7783-20-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Sulfuric Acid Diammonium Salt
IUPAC Name water
SMILES O

Specifications

Boiling Point 103°C
Color Colorless
Physical Form Liquid
Quantity 20 L
Grade Reagent
Packaging Cubitainer
Specific Gravity 1.2189
Chemical Name or Material Ammonium Sulfate, 3.5 Molar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.