Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Sulfate, 99.0% (ACS Guaranteed Reagent), MilliporeSigma™
GSA_VA

Catalog No. MAX13851 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAX13851 500 g
MAX13853 2.5 kg
MAX13855 12 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MAX13851 Supplier MilliporeSigma Supplier No. AX13851
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
PubChem CID 6097028
ChEBI CHEBI:62946
IUPAC Name sulfuric acid diamine
SMILES N.N.OS(O)(=O)=O

Specifications

Physical Form Crystalline
Quantity 500 g
Linear Formula (NH4)2SO4
Formula Weight 132.14
Percent Purity ≥99.0%
Grade ACS
Density 1.77g/cm3 (20°C); Bulk Density: 850 kg/m3
Chemical Name or Material Ammonium Sulfate, 99.0%, Suprapur™
Recommended Storage Store without restrictions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.